Anti-MUC1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-CW0
75930-156EA 635.22 USD
75930-156
Anti-MUC1 Rabbit Polyclonal Antibody
Antibodies
Mucin 1 (MUC1) is membrane-bound protein member of the mucin family. Mucins are O-glycosylated proteins that play an essential role in forming protective mucous barriers on epithelial surfaces and also play a role in intracellular signaling (1). MUC1 is expressed on the apical surface of epithelial cells that line the mucosal surfaces of many different tissues including lung, breast stomach and pancreas. MUC1 is proteolytically cleaved into alpha and beta subunits that form a heterodimeric complex. The N-terminal alpha subunit functions in cell-adhesion and the C-terminal beta subunit is involved in cell signaling. Overexpression, aberrant intracellular localization, and changes in glycosylation of this protein have been associated with carcinomas (1).

Anti-MUC1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 122 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: MUC1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR