Anti-IgG1 Goat Mouse Polyclonal Antibody (TRITC)

Supplier: Rockland Immunochemical
610-100-040
75930-136EA 258.06 USD
75930-136
Anti-IgG1 Goat Mouse Polyclonal Antibody (TRITC)
Antibodies
Anti-Mouse IgG1 Rhodamine Antibody generated in goat detects reactivity to Mouse IgG1 (Gamma 1 chain). Secreted as part of the adaptive immune response by plasma B cells, immunoglobulin G constitutes 75% of serum immunoglobulins. IgG1 chain constitutes 66% of the IgG subclass and has a high affinity for binding to the Fc receptor of phagocytic cells. Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product, consideration must be given to species and immunoglobulin specificity, conjugate type, fragment and chain specificity, level of cross-reactivity, and host-species source and fragment composition.

Mouse IgG1 Antibody Rhodamine Conjugated is designed for immunofluorescence microscopy, fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis, including multicolor imaging, utilizing various commercial platforms.

Type: Secondary
Antigen: IgG1
Clonality: Polyclonal
Clone:
Conjugation: TRITC
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR