Anti-MECP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-X14
75929-924EA 656.12 USD
75929-924
Anti-MECP2 Rabbit Polyclonal Antibody
Antibodies
MeCP2 (Methyl-CpG Binding Protein 2) is a chromosomal protein that binds to methylated DNA. It can bind specifically to a single methyl-CpG pair and is not influenced by sequences flanking the methyl-CpGs. MeCP2 has been shown to mediate transcriptional repression through interaction with histone deacetylase and the corepressor SIN3A. Defects in MeCP2 are the cause of Rett syndrome (RTT). RTT is an X-linked dominant disease; it is a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females. Recent studies have reported a new phosphorylation site at Ser421. Phosphorylation and dephosphorylation of this site may be involved in regulation of behavioral responses to chronic antidepressant treatment. MeCP2 pS421 Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Ser421 MeCP2 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~55 kDa truncated MeCP2 protein phosphorylated at Ser421. Immunolabeling of the MeCP2 band is blocked by preadsorption with the phospho-peptide used as antigen but not by the corresponding dephospho-peptide. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR