Anti-ME2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-CS9
75929-920EA 635.22 USD
75929-920
Anti-ME2 Rabbit Polyclonal Antibody
Antibodies
ME2 is a homotetrameric, mitochondrial NAD-dependent malic enzyme that catalyzes the oxidative decarboxylation of malate to pyruvate (1). It is related to malic enzyme 1 (ME1), a cytoplasmic NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis (2). The expression of both malic enzymes is reciprocally regulated by p53; this regulation has been shown to modulate metabolism and senescence (3). Certain single-nucleotide polymorphism haplotypes of the ME2 gene have been shown to increase the risk for idiopathic generalized epilepsy (4).

Anti-ME2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 65 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ME2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR