MDA-MB-468 Whole Cell Lysate, Rockland Immunochemicals
Supplier: Rockland Immunochemical
W09-001-GG9
75929-904EA 174.48 USD
75929-904
MDA-MB-468 Whole Cell Lysate, Rockland Immunochemicals
Cells
MDA-MB-468 Whole Cell Lysate
  • Ready-to-use lysates
  • positive controls
Ready-to-use whole cell lysates produced by Rockland Immunochemicals are derived from cell lines or tissues using highly refined extraction protocols to ensure exceptionally high quality, protein integrity and lot-to-lot reproducibility.  All extracts are tested by SDS-PAGE using 4-20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Ready-to-use lysates are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Lysates are prepared in denaturing buffer WITHOUT dissociating agents (i.e. no 2-mercaptoethanol or dithiothreitol has been added). Heat lysate to 95° C for 5 minutes and rapidly cool.  If dissociating conditions are desired, add reducing agent prior to heating.  The recommended loading volume per lane is 10-20 µl depending on the size format of your gel.