Anti-MAP1LC3A Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-CQ4
75929-848EA 635.22 USD
75929-848
Anti-MAP1LC3A Rabbit Polyclonal Antibody
Antibodies
Microtubule-associated proteins (MAPs) regulate microtubule stability and play critical roles in neuronal development and plasticity (1). MAP1LC3 is a subfamily of three related proteins belonging to the MAP1 LC3 family and it includes MAP1LC3A, MAP1LC3B, and MAP1LC3C (2). MAP1LC3 is the mammalian homolog of yeast ATG8 and is essential for autophagy and associated with the autophagosome membranes after processing (3). The three isoforms exhibit distinct expression patterns and both MAP1LC3A and MAP1LC3B but not MAP1LC3B, are post-translationally modified, suggesting the three isoforms may have different physiological functions (4).

Anti-MAP1LC3 Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 14 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: MAP1LC3A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR