Anti-LMNA Mouse Monoclonal Antibody [Clone: 4C4]

Supplier: Rockland Immunochemical
200-301-X12
75929-514EA 659.25 USD
75929-514
Anti-LMNA Mouse Monoclonal Antibody [Clone: 4C4]
Antibodies
Lamins A and C are nuclear structural proteins that are part of the intermediate filament family and coded for by the same gene (LMNA). Lamins A and C are nearly identical except for their carboxy termini. Mutations in the gene encoding lamins A/C have been shown to cause a variety of diseases including autosomal dominant Emery-Dreifuss muscular dystrophy, autosomal dominant limb-girdle muscular dystrophy and Charcot-Marie-Tooth disorder type 2. Lamin A/C Antibody is ideal for researchers interested in cancer research.

Anti-Lamin A/C Antibody is suitable for Western Blots and Immunofluorescence, and it is specific for the ~64 and 74 kDa lamin A and C proteins. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: LMNA
Clonality: Monoclonal
Clone: 4C4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR