Anti-KREMEN2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-CG8
75929-480EA 635.22 USD
75929-480
Anti-KREMEN2 Rabbit Polyclonal Antibody
Antibodies
Kremen (Kringle containing protein marking the eye and the nose) proteins are type I transmembrane proteins that contain extracellular kringle, WSC and CUB domains and an intracellular region without any conserved motifs. Kremens bind a subset of the secreted Dickkopf proteins (Dkk 1, 2, and 4) with high affinity to modulate the canonical Wnt signaling pathway that is transduced by the ternary receptor complex composed of Wnt, Frizzled, and the LDL receptor related protein 5/6 (LRP5/6) co-receptor. Recent experiments have shown that KREMEN2 is a regulator of bone remodeling and raise the possibility that antagonizing KREMEN2 might prove beneficial in patients with bone loss disorders.

Anti-KREMEN2 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 49 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: KREMEN2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR