Anti-Kdm5b Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
100-401-V92
75929-274EA 520.3 USD
75929-274
Anti-Kdm5b Rabbit Polyclonal Antibody
Antibodies
JARID1B is a histone demethylase which specifically demethylates mono-, di, and trimethylated lysine 4 of histone H3. It acts as a transcriptional repressor for the tumor suppressor genes BRCA1 and HOXA5 and has been associated with breast cancer development. In contrast, in melanoma JARID1B itself may function as a tumor suppressor. Anti-Jarid1b Antibody is ideal for research in Epigenetics, Chromatin Remodeling and Cancer.

Anti-Jarid1b Antibody is suitable for ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 175 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: Kdm5b
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR