Anti-SUC1 Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemical
200-106-189-0100
75929-184EA 531.79 USD
75929-184
Anti-SUC1 Goat Polyclonal Antibody (BAC)
Antibodies
Anti-Invertase antibody recognizes invertase which hydrolyzes the alpha-1,4-glycosidic bond between alpha-D-glucose and beta-D-fructose of sucrose. Bees use invertase to convert nectar to honey. In the confectionary industry, yeast-derived invertase is used to hydrolyze sucrose to glucose and fructose as fructose is more sweeter and less prone to crystallization.

Anti-Invertase antibody is ideal for investigators interested in Metabolism and Cell Biology.

Anti-Invertase antibody has been assayed against 1.0 ug of Invertase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:4,000 to 1:20,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: SUC1
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR