Human Recombinant IL-21 (from E. coli)
Supplier: Rockland Immunochemical
009-F01-V61-0010 009-F01-V61-0100
75929-124EA 8882 USD
75929-124 75929-126 75929-128 75929-130
Human Recombinant IL-21 (from E. coli)
Proteins and Peptides
Recombinant Human IL-21 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-21 (IL-21) is a member of the common gamma-chain family of cytokines (includes IL-2, IL-4, IL-7, IL-9, IL-13, and IL-15) and is expressed by activated CD4+ T cells. IL-21 has a private receptor known as IL-21R, which is expressed on T, B, and NK cells. Within the B cell lineage, IL-21 regulates IgG1 production and cooperates with IL-4 for the production of multiple Ab classes in vivo. Initial studies have demonstrated that IL-21 has pleiotropic effects on the proliferation, differentiation, and effector functions of B, T, natural killer, and dendritic cells. Recombinant human IL-21 is a non-glycosylated protein, containing 133 amino acids, with a total molecular weight of 15.5 kDa.
Interleukin-21 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-21 in immunological assays.