Mouse Recombinant IL-11
Supplier: Rockland Immunochemical
010-F01-V49-0010 010-F01-V49-0100
75929-022EA 8882 USD
75929-022 75929-024 75929-026 75929-028
Mouse Recombinant IL-11
Proteins and Peptides
Recombinant Mouse IL-11 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin 11 (IL-11) is a pleotropic cytokine closely related to IL-6 in function, but is overall one of the lesser characterized interleukins. In hematopoietic cell populations, IL-11 is thought to induce megakaryocyte differentiation. In non-hematopoietic populations, IL-11 is thought to be able to stimulate hepatic acute-phase proteins. Human IL-11 can stimulate murine cells. Recombinant mouse IL-11 is a non-glycosylated protein, containing 179 amino acids, with a molecular weight of 19.3 kDa.
Interleukin-11 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-11 in immunological assays.