Human Recombinant INF gamma
Supplier: Rockland Immunochemical
009-001-V20-0100 009-001-V20-0020
75928-984EA 1819 USD
75928-984 75928-986 75928-988
Human Recombinant INF gamma
Proteins and Peptides
Recombinant Human INF gamma control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interferon gamma, IFNγ, is a major immune-modulating molecule produced mainly by T cells and natural killer cells activated by antigens, mitogens or alloantigens. Most immune cells express IFN-γ receptors and respond to IFNγ-induced signaling by up-regulating MHC class I expression. Human IFNγ does not show cross-reactivity with mouse. Recombinant human IFNγ is a non-glycosylated protein, containing 144 amino acids, with a molecular weight of 16.88 kDa.
Interferon-gamma Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interferon-gamma in immunological assays.