Human Recombinant INF alpha 2a
Supplier: Rockland Immunochemical
009-001-V18-0100 009-001-V18-0020
75928-972EA 1640 USD
75928-972 75928-974 75928-976
Human Recombinant INF alpha 2a
Proteins and Peptides
Recombinant Human INF alpha 2a control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interferon-alpha 2a (IFNα 2a) is a Type I interferon made by leukocytes during viral infection. It has anti-viral and non-proliferative activity induced by JAK1-STAT signaling. IFN-alpha 2s are the most widely used type for therapeutic purposes. IFNα 2b is a variant of IFNα 2a that differs by one amino acid. Recombinant human IFNα 2a is a non-glycosylated protein, containing 166 amino acids, with a molecular weight of 19.37 kDa.
Interferon-alpha 2a Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interferon-alpha 2a in immunological assays.