Anti-IL36RN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-BX6
75928-922EA 635.22 USD
75928-922
Anti-IL36RN Rabbit Polyclonal Antibody
Antibodies
IL-36RN is a member of the interleukin 1 cytokine family whose gene and eight other interleukin 1 family genes form a cytokine gene cluster on chromosome 2 (1). IL-36RN specifically inhibits the activation of NF-kappaB induced IL-36A (2). The opposing activities of these cytokines have been suggested to be involved in the regulation of skin inflammation (3). Like the IL-36 cytokines (IL-36A, IL-36B and IL-36G), IL-36RN requires post-translational processing for full agonist activity, but the cleavage mechanism is currently unknown (4).

Anti-IL-36RN Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 17 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: IL36RN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR