Anti-IL22 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-BV7
75928-880EA 635.22 USD
75928-880
Anti-IL22 Rabbit Polyclonal Antibody
Antibodies
Interleukin-22 (IL-22) is a cytokine important for the modulation of tissue responses during inflammation (1). Unlike the distantly related IL-10, IL-22 does not inhibit the production of proinflammatory cytokines in monocytes in response to LPS, but it has some inhibitory effects on IL-4 production from Th2 T cells. IL-22 is expressed by both the adaptive arm of the immune system such as CD4 T cell subsets including Th17 cells, as well as by innate lymphocytes such as NK and LTi-like cells (2). IL-22 is highly expressed in several chronic inflammatory conditions, and studies suggest that IL-22 plays both inflammatory and protective roles (3).

Anti-IL-22 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 20 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: IL22
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR