Anti-IL21 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemical
600-401-G98
75928-874EA 635.22 USD
75928-874
Anti-IL21 Rabbit Polyclonal Antibody
Antibodies
IL-21 antibody detects human IL-21 precursor. A novel cytokine was recently identified in human and mouse and designated IL-21 , which has significant homology to IL-2, IL-4, and IL-15. The receptor for IL-21 (IL-21R, also termed NILR for novel Interleukin receptor) is a new member of the class I cytokine receptor family. IL-21R forms a complex with the common cytokine receptor gamma chain, gammac, and mediates IL-21 signaling. IL-21 and its receptor activate JAK-STAT signaling pathway. IL-21 is expressed in activated T cells, and HL-60 and THP-1 cell lines. IL-21 plays a role in the proliferation and maturation of NK, B and T cell populations. Anti-IL-21 antibodies are ideal for investigators involved in Cytokines and Growth factors research.

Anti-IL-21 Antibody is suitable for use in E,WB, and ICC. Expect a band approximately ~17.9kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: IL21
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR