Human Serum (Lyophilized), Rockland Immunochemicals
Supplier: Rockland Immunochemical
D314-05
75928-834EA 277 USD
75928-834
Human Serum (Lyophilized), Rockland Immunochemicals
Sera (Serum)
Lyophilized Non-Sterile Serum
  • prepared by removal of fibrinogen through clot formation
  • separated from cellular components and purified through centrifugation and filtration techniques
Human Serum is used as a component of bioassays, immunoassays or enzyme assays. Human Serum provides a broad spectrum of macromolecules, carrier proteins for lipoid substances and trace elements, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors that promote cell growth and health. Human serum is available as a sterile preparation for use as a cell culture supplement.
Human serum is suitable for use as a control or diluent in assays and most other immunological methods.