Human Recombinant IL-22
Supplier: Rockland Immunochemical
009-001-J33-0010 009-001-J33-0100
75928-796EA 8070 USD
75928-796 75928-798 75928-800 75928-802
Human Recombinant IL-22
Proteins and Peptides
rHuman IL-22 control protein
  • Control Protein
  • PCA Assays
  • ImmunoBlot
Interleukin-22 (IL-22), also called IL-TIF, is an IL-10 family member, produced by activated dendritic cells and T lymphocytes. IL-22 signals via the IL-22R and IL-10Rbeta receptors to act as a potent mediator of the cellular inflammatory response. Recombinant human IL-22 is a non-disulfide-linked homodimer, containing two 146 amino acids, with a total molecular weight of 33.6 kDa.
Interleukin-22 Recombinant Protein is suitable as a control for polyclonal or monoclonal anti-Interleukin-22 in immunological assays.