HSPA1AB
Supplier: Rockland Immunochemical
009-001-F63S
75928-744EA 380.3 USD
75928-744 75928-748
HSPA1AB
Proteins and Peptides
HSP70 control protein-GST fusion
  • Control Protein
  • PCA Assays
  • ImmunoBlot
HSP70 is a member of the heat shock protein family and is synthesized by cells of many organisms in response to stress (1). HSP70 is found mostly but not exclusively in the nucleus of unstressed cells. For several hours after a short heat shock, however, it is strongly concentrated in nucleoli (2). Nucleoli are transiently damaged by such a heat shock: their morphology changes and assembly and export of ribosomes is blocked for several hours. HSP70 helps to stabilize the morphological changes of the nucleoli. HSP70 Protein is ideal for investigators involved in Signaling Proteins, Cell Stress & Chaperone Proteins, Apoptosis/Autophagy, Cancer, Cardiovascular Disease, Cellular Stress, and Inflammation research.
HSP70 Protein is stored in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 0.25mM DTT, 0.1mM PMSF, 25% glycerol. HSP70 Protein is suitable for use in Western Blot and ATPase Assay. Expect a band approximately ~96kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.