Horse Gamma Globulin Fraction
Supplier: Rockland Immunochemical
D607-1000 D607-0100
75928-524EA 155.67 USD
75928-524 75928-526
Horse Gamma Globulin Fraction
Antibodies
Lyophilized Gamma Globulin Fraction
  • prepared from normal serum by a multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
  • control reagent
  • Western Blotting and ELISA experiments
Horse Gamma Globulin Fraction consists of the fraction of blood serum which contains whole antibodies as well as other non-albumin plasma proteins. Gamma globulins can be utilized therapeutically to temporarily boost an patient's immunity (such as after an immunosuppressive infection) or to increase the likelihood of kidney transplant acceptance. Gamma Globulin Fraction can be utilized in molecular biology experiments as a control reagent.
Horse Gamma Globulin Fraction can be utilized in Western Blotting and ELISA experiments.