Hamster Brain (Cerebellum), Rockland Immunochemicals
Supplier: Rockland Immunochemical
HM-T152
75928-356EA 78.36 USD
75928-356
Hamster Brain (Cerebellum), Rockland Immunochemicals
Specimens Preserved Specimens Preserved Animal Materials
Hamster Brain (Cerebellum)
  • Harvested Animal Tissue
Fresh-Frozen collection of tissue
healthy non-immunized animals
useful for staining assays, controls, lysates, sequencing