Thin-layer chromatography plates (TLC), SiliaPlate™ TLC plate, Coating material: Silica Gel, Backing Material: Plastic, Fluorescent Indicator: F254, Plate size: 200×200 mm, Layer thickness: 200 µm
Supplier: SiliCycle
Thin-layer chromatography plates (TLC), SiliaPlate™ TLC plate, Coating material: Silica Gel, Backing Material: Plastic, Fluorescent Indicator: F254, Plate size: 200×200 mm, Layer thickness: 200 µm