High performance thin-layer chromatography plates (HPTLC), SiliaPlate™ HPTLC plate, Coating material: RP-8 (C8) Modified Silica Gel, Backing Material: Glass, Fluorescent Indicator: F254, Plate size: 100×100 mm, Layer thickness: 150 µm
Supplier: SiliCycle
High performance thin-layer chromatography plates (HPTLC), SiliaPlate™ HPTLC plate, Coating material: RP-8 (C8) Modified Silica Gel, Backing Material: Glass, Fluorescent Indicator: F254, Plate size: 100×100 mm, Layer thickness: 150 µm