Rotopic™ Replacement Vacuum Pump, Excelta®
Supplier: Excelta
2000V
75880-670EA 645.86 USD
75880-670
Rotopic™ Replacement Vacuum Pump, Excelta®
Pumps Vacuum Pumps
The 2000V is a Rotopic™ replacement vacuum pump.
  • Pump generates up to 10" of mercury
  • Supplied complete with a 1/8" adaptor