KWIK-STIK 2 Pack Proteus mirabilis ATCC® 29906™*, Microbiologics
Supplier: MICROBIOLOGICS, INC.
01215P
75879-060EA 100.99 USD
75879-060
KWIK-STIK 2 Pack Proteus mirabilis ATCC® 29906™*, Microbiologics
Controls and Calibration Verification Materials
Everything needed to grow reference cultures for QC testing is included in this one handy device.
  • Over 900 strains available
  • Ready-to-use format saves time and money
  • Refrigerated storage is easy and economical
  • Online Certificate of Analysis provides detailed strain information
  • Traceability to reference cultures ensures authenticity
  • Technical Support experts available for guidance
All-in-one design reduces the risk of contamination and strains are less than or equal to four passages from reference culture.
Available in a two pack, universally used for all types of microbiological control testing.
Delivery information: KWIK-STIK’s simple, all-inclusive design makes life easier for lab techs while reducing chances for errors. Each of it contains a qualitative lyophilized microorganism pellet, ampoule of hydrating fluid, and inoculating swab.