Sulfate Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS MS
For quantitative determination of sulfate ion and evaluation of drug effects on sulfate metabolism
- Sensitive and accurate. Detection range 0.02 mM (0.19 mg/dL) to 2 mM (19.2 mg/dL) sulfate in 96-well plate assay.
- Simple and high-throughput. The procedure involves addition of a single working reagent and incubation for 5 min.
- Method: OD600nm
- Samples: Serum, urine, food and environment
INORGANIC SULFATE is one of the most abundant anions in mammalian plasma. Sulfate plays important physiological roles in activating and detoxifying xenobiotics, steroids, neurotransmitters, and bile acids. Sulfate is needed for the biosynthesis of glycosaminoglycans, cerebroside sulfate, and heparin sulfate. Undersulfation of cartilage proteoglycans has been associated with human inherited osteochon-drodysplasia disorders. In mammals, sulfate homeostasis is regulated by the kidney. The majority of filtered sulfate is absorbed in the proximal tubules, and only 5–20% of the filtered load is excreted into the urine. Simple, direct and automation-ready procedures for quantitative determination of inorganic sulfate find wide applications in research and drug discovery. BioAssay Systems’ sulfate assay kit is designed to measure sulfate concentration in biological fluids such as serum and urine. The improved method utilizes the quantitative formation of insoluble barium sulfate in polyethylene glycol. The turbidity measured between 540 and 610nm is proportional to sulfate level in the sample.