EnzyChrom™ Lactate Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
ECLC-100
75878-070EA 667.1 USD
75878-070
EnzyChrom™ Lactate Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative colorimertic determination of L-lactate (L-lactic acid) and evaluation of drug effects on its metabolism.
  • Linear detection range 0.05 - 2 mM L-Lactate in 96-well plate assay
  • Convenient room temperature assay. No 37°C heater is needed
  • Can be readily automated to process thousands of samples per day
Lactate is generated by lactate dehydrogenase (LDH) under hypoxic or anaerobic conditions. Monitoring lactate levels is, therefore, a good indicator of the balance between tissue oxygen demand and utilization and is useful when studying cellular and animal physiology. Simple, direct and automation-ready. Procedures for measuring lactate concentration are very desirable. BioAssay Systems’ EnzyChrom™ lactate assay kit is based on lactate dehydrogenase catalyzed oxidation of lactate, in which the formed NADH reduces a formazan (MTT) reagent. The intensity of the product color, measured at 565 nm, is proportionate to the lactate concentration in the sample.