EnzyChrom™ Citrate Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
ECIT-100
75878-068EA 642.36 USD
75878-068
EnzyChrom™ Citrate Assay Kit, BioAssay Systems
Assays Cellular Assays
For quantitative determination of citrate or citric acid and evaluation of drug effects on citrate metabolism.
  • Linear detection range 4 to 400 μM citrate for colorimetric assays and 0.5 to 40 μM for fluorimetric assays
  • Can be readily automated to process thousands of samples per day
Citrate is an intermediate in the citric acid cycle and is involved in fatty acid synthesis. BioAssay Systems’ Citrate Assay Kit provides a simple, and automation-ready procedure for measuring citrate concentration. Citrate is converted into pyruvate which is then oxidized with the conversion of the dye into a colored and fluorescent form. The color intensity at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the citrate concentration in the sample.