EnzyChrom™ Amylase Assay Kit, BioAssay Systems
Supplier: BIOASSAY SYSTEMS
ECAM-100
75878-054EA 665.1 USD
75878-054
EnzyChrom™ Amylase Assay Kit, BioAssay Systems
Assays Enzyme Assays
For quantitative determination of alpha-amylase activity.
  • Linear detection range 0.3 to 50 U/L α-amylase in 96-well plate assay
Amylase belongs to the family of glycoside hydrolase enzymes that break down starch into glucose molecules by acting on α-1,4-glycosidic bonds
BioAssay Systems’ EnzyChrom™ α-amylase assay method involves two steps: (1). α-amylase in the sample hydrolyzes starch and the product is rapidly converted to glucose by α-glucosidase and hydrogen peroxide by glucose oxidase; (2). hydrogen peroxide concentration is determined with a colorimetric reagent.
The α-amylases (EC 3.2.1.1) cleave at random locations on the starch chain, ultimately yielding maltotriose and maltose, glucose and "limit dextrin" from amylose and amylopectin. In mammals, α-amylase is a major digestive enzyme. Increased enzyme levels in humans are associated with salivary trauma, mumps due to inflammation of the salivary glands, pancreatitis and renal failure. Simple, direct and automation-ready. Procedures for measuring amylase activity are very desirable.