PTFE Rotary Evaporator Vial Adapter, Ace Glass Incorporated
Supplier: Ace Glass
13290-12 13290-122
75877-616EA 53.84 USD
75877-616 75877-706
PTFE Rotary Evaporator Vial Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
PTFE connecting adapter for connection of GPI threaded vials to a rotary evaporator vapor tube or trap.
  • Adapter made of PTFE
  • Suitable for vacuum work
  • Includes either a FETFE or CHEMRAZ O-Ring
  • CHEMRAZ O-Ring should be used with Methylene Chloride or Acetone