Five-Neck Round-Bottom Flask, Ace Glass
Supplier: Ace Glass
6957-65
75877-634EA 6256.37 USD
75877-634
Five-Neck Round-Bottom Flask, Ace Glass
Flasks Round Bottom Flasks
Large round bottom flask with five necks, including one neck with an o-ring groove flange.
  • High quality borosilicate glass
  • Flask has five necks arranged 3 in-line, and 2 that are 90 degrees front and back of the in-line necks
  • All necks are vertical
Five neck round bottom flask with one center and three side necks having 45/50 joints, and one side neck having a 60 mm o-ring groove flange.