Toxoplasma sp., Animal IHC Positive Control Slides, Newcomer Supply

Supplier: Newcomer Supply
3801A 3801B
75877-138BX 91.27 USD
75877-138 75877-140
Toxoplasma sp., Animal IHC Positive Control Slides, Newcomer Supply
Slides Microscopy Control Slides
Toxoplasma sp., animal control slides contain sections of positive staining rat organ and negative staining human myometrium.

  • Tissue type: (+) Rat Organ / (-) Myometrium
  • All control tissue is sectioned on Superfrost® Plus slides
  • Staining validation technical memo included in each set
  • All tissues are formalin fixed, paraffin embedded FFPE
  • All slides are air dried
  • IHC slides are cut at 4 µm
  • Standard tissue placement is mid-high on slide

Newcomer supply Toxoplasma sp., Animal Control Slides are for the positive immunohistochemical staining of Toxoplasma, a crescent shaped sporozoan found as an intracellular parasite in various tissues.

Staining Results:
Toxoplasma positive expression will stain golden brown/brown organisms.
Myometrium will stain negative.
Nuclei will stain blue.

These positive control slides are intended to be used to verify histological techniques and reagent reactivity. These slides are to be used for the qualitative purpose of determining positive or negative results, and are not intended to be used for any quantitative purpose.

Ordering information: All controls are cut in serial sections.
10 set = 10 unstained slides and 1 stained slide.
98 set = 98 unstained slides and 2 stained slides. (First & Last Slides are Stained)
Reactivity of IHC control slides are guaranteed for one year from the date of receipt and should be revalidated after one year to verify continued reactivity.
Please note the following: Allow 2-3 weeks for delivery for large sets. In human tissue unless otherwise indicated.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR