LaboPlast Stand for Imhoff Sedimentation Funnels, Bürkle
Supplier: BURKLE INC
7118-2000
75876-084EA 191.49 USD
75876-084
LaboPlast Stand for Imhoff Sedimentation Funnels, Bürkle
Support Stands
The stand for sedimentation funnels is used to keep, hold or store Imhoff funnels safely and reliably during sedimentation.
  • Adjustable height
  • For funnels with Ø 90 mm (3.54") or more
Two funnel holders ensure that Imhoff funnels will be held safely and securely and can be placed in an absolutely vertical position. Stand with two holders for sedimentation funnels. Innovative magnet concept, no clamping screws are necessary.