TechniCloth® Dry Wipers, Sterile, Texwipe®
Supplier: Texwipe
STX609
75875-934BG 39.48 USD
75875-934
TechniCloth® Dry Wipers, Sterile, Texwipe®
Wipes Cleanroom Wipes Sterile Cleanroom Wipes
TechniCloth® provides the lowest level of releasable ions and non-volatile contaminants. It is made from 45% polyester and 55% cellulose to provide the optimum balance of sorption, durability, and cleanliness.
  • Excellent chemical resistance for compatibility with a variety of solutions
  • Designed for spill control, wiping, and cleaning equipment and parts surfaces
  • Also useful for applying and removing lubricants, adhesives, and residues
  • Wipes will clean sterile environments and dispense solutions
Each package is gamma irradiated to 10⁻⁶ Sterility Assurance Level.