Eppendorf® twin.tec® 96- & 384-Well Microbiology PCR Plates
Supplier: Eppendorf
0030129300 0030129318 0030129326 0030129334 0030129342 0030129350
75874-684CS 120.57 USD
75874-684 75874-686 75874-688 75874-690 75874-692 75874-694
Eppendorf® twin.tec® 96- & 384-Well Microbiology PCR Plates
Microplates PCR Plates
Eppendorf® Microbiology PCR plates combine extremely thin walled polypropylene wells for optimal heat transfer to the sample and extremely robust polycarbonate frames for ultimate rigidity and torque-resistance
These twin.tec microbiology plates are sterile, individually blister packed, and certified absent of bacterial DNA. Available in 96- or 384-well formats.
Packaging: Plates are supplied individually wrapped, in a case of 10.