Antigen Retrieval Reagent, pH 6 (10X), Enzo Life Sciences
Supplier: Enzo Life Sciences
ACC112-0100
75873-994EA
172.22
USD
75873-994
Antigen Retrieval Reagent, pH 6 (10X), Enzo Life Sciences
Protein Extraction
Protein Extraction Kits
Antigen Retrieval Reagent, pH 6 (10X) is used to recover antigens masked by fixation in cross linking fixatives such as formalin.