Tetrahydrofolic acid

Supplier: Spectrum Chemical Mfg. Corp.

T2824-1GM
75873-506EA 386.84 USD
75873-506 75873-508 75873-510
Tetrahydrofolic acid
Tetrahydrofolic acid
Formula: C₁₉H₂₃N₇O₆
Storage Temperature: Freezer
MDL Number: MFCD00135583
CAS Number: 135-16-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR