3,3',5-Triiodo-L-thyronine free acid

Supplier: Spectrum Chemical Mfg. Corp.

T2780-100MG
75873-500EA 105.15 USD
75873-500
3,3',5-Triiodo-L-thyronine free acid
3,3',5-Triiodo-L-thyronine

3,3',5-Triiodo-L-thyronine, Free Acid, also known as Triiodothyronine, is a thyroid hormone which can be used as ingredients in certain fat-loss supplements. This product affects almost every physiological process in the body, including growth and development, metabolism, body temperature, and heart rate.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₅H₁₂I₃NO₄
Storage Temperature: Freezer
MDL Number: MFCD00002593
CAS Number: 6893-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR