3-Nitrobenzaldehyde

Supplier: Spectrum Chemical Mfg. Corp.

N2160-500GM
75872-440EA 54.14 USD
75872-440 75873-090
3-Nitrobenzaldehyde
3-Nitrobenzaldehyde

3-Nitrobenzaldehyde, also known as meta-nitrobenzaldehyde, is an organic aromatic compound obtained via the mono-nitration of benzaldehyde with nitric acid.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₅NO₃
Storage Temperature: Ambient
MDL Number: MFCD00007249
CAS Number: 99-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR