Nonanophenone

Supplier: Spectrum Chemical Mfg. Corp.

N1544-25ML
75873-084EA 188.55 USD
75873-084
Nonanophenone
Nonanophenone
Formula: C₆H₅C(O)(CH₂)₇CH₃
Storage Temperature: Ambient
MDL Number: MFCD00048965
CAS Number: 6008-36-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=954329dfa1dfb3703f537ff434c29d3d07d6dc94&vid=8404e060bb250a6a8e0cec57e16cf8c5c1d53d5a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR