3-Hydroxy-p-anisaldehyde

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Isovanillin, Isovanicaline, 3-Hydroxy-4-methoxybenzaldehyde, Isovanillic aldehyde

I3248-100GM I3248-25GM
75872-822EA 297.1 USD
75872-822 75872-824
3-Hydroxy-p-anisaldehyde
3-Hydroxy-p-anisaldehyde
Formula: C₈H₈O₃
Storage Temperature: Ambient
MDL Number: MFCD00003369
CAS Number: 621-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR