Fluorescein disodium salt

Supplier: Spectrum Chemical Mfg. Corp.

F1272-100GM F1272-25GM F1272-500GM
75872-508EA 93.51 USD
75872-508 75872-510 75872-512
Fluorescein disodium salt
Fluorescein disodium salt
Formula: C₂₀H₁₀Na₂O₅
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR