EDTA (ethylenediamine tetraacetic acid)

Supplier: Spectrum Chemical Mfg. Corp.

E3532-125GM E3532-2.5KG E3532-500GM E3532-12KG E3532-45KGBL
75872-496EA 69.2 USD
75872-496 75872-498 75872-500 76014-446 76014-448
EDTA (ethylenediamine tetraacetic acid)
EDTA (Ethylenediamine tetraacetic acid)

(Ethylenedinitrilo)tetraacetic Acid, Powder is generally abbreviated as EDTA, a colorless, water soluble, aminopolycarboxylic acid and solid. It's used for its chelating properties and as a ligand. It is used by many industries for application ranging from sequestering agent to chelation therapy. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for consumption.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₀H₁₆N₂O₈
Storage Temperature: Ambient
MDL Number: MFCD00003541
CAS Number: 60-00-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ac89cc787258fff61bfc9612051bdc09d2233&vid=adf1cfd608e336342a4765f47271a739c6c17113&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR