epi-Testosterone

Supplier: Spectrum Chemical Mfg. Corp.

E3287-100MG
75872-492EA 327.82 USD
75872-492
epi-Testosterone
epi-Testosterone

Epitestosterone is an androstanoid that is the C-17 epimer of testosterone. Non-graded Epitestosterone supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₉H₂₈O₂
Storage Temperature: Ambient
MDL Number: MFCD00067129
CAS Number: 481-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR