3,4-Diaminotoluene

Supplier: Spectrum Chemical Mfg. Corp.

D1232-100GM
75872-220EA 290.35 USD
75872-220 75872-222
3,4-Diaminotoluene
3,4-Diaminotoluene

3,4-Diaminotoluene is used as an epoxy resin curing agent, comonomer for PU polyols and a colorant in hair dyes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD00007728
CAS Number: 496-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR