Chlorohydroquinone

Supplier: Spectrum Chemical Mfg. Corp.

C2211-100GM C2211-25GM
75872-144EA 244.2 USD
75872-144 75872-146
Chlorohydroquinone
Chlorohydroquinone
Formula: C₆H₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00002341
CAS Number: 615-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR