sec-Butyl-4-hydroxybenzoate

Supplier: Spectrum Chemical Mfg. Corp.

B1533-100GM B1533-25GM
75872-002EA 487.05 USD
75872-002 75872-004
sec-Butyl-4-hydroxybenzoate
sec-Butyl 4-hydroxybenzoate
Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00059626
CAS Number: 17696-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR