(±)-2-Butanol
Supplier: Spectrum Chemical Mfg. Corp.
sec-Butyl Alcohol is also known as 2-Butyl alcohol and sec-Butanol. It is used as a solvent and can be converted to butanone which is an industrial solvent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
H₃CCH₂CH(OH)CH₃ Density: 0.808 Storage Temperature: Ambient |
MDL Number:
MFCD00004569 CAS Number: 78-92-2 UN: 1120 ADR: 3,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...