Ethyl hydrogen adipate

Supplier: Spectrum Chemical Mfg. Corp.

A2023-100GM A2023-25GM
75871-806EA 291.7 USD
75871-806 75871-808
Ethyl hydrogen adipate
Ethyl hydrogen adipate
Formula: C₈H₁₄O₄
Density: 0.98
Storage Temperature: Ambient
MDL Number: MFCD00004419
CAS Number: 626-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR